p-Toluidinium chloride ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America
Danger

Synonyms: p-Toluidine hydrochloride, 4-Toluidine HCl, p-Toluidine HCl, 4-Aminotoluene hydrochloride, 4-Methylaniline HCl, 4-Methylaniline hydrochloride, 4-Aminotoluene HCl, 4-Toluidine hydrochloride

T0303-500G T0303-25G
TCT0303-500GEA 153 USD
TCT0303-500G TCT0303-025G
p-Toluidinium chloride ≥98.0% (by HPLC, total nitrogen)
p-Toluidinium chloride

CAS Number: 540-23-8
MDL Number: MFCD00035497
Molecular Formula: C7H9N
Molecular Weight: 143.61
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Color: White
Melting point (°C): 243

Formula: C₇H₁₀ClN
MW: 143.62 g/mol
Melting Pt: 243 °C
Density: 1.19 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00035497
CAS Number: 540-23-8
EINECS: 208-740-8
UN: 2811
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=567bcf87dedf28bc79bb819bd83eea64a94bcef3&vid=567bcf87dedf28bc79bb819bd83eea64a94bcef3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC,N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR