Ammonium thioglycolate ∼60% in water

Supplier: TCI America

Synonyms: Ammonium thioglycolate solution, Thioglycolic Acid Ammonium Salt, Ammonium Mercaptoacetate

A2217-25G A2217-500G
TCA2217-25GEA 20 USD
TCA2217-25G TCA2217-500G
Ammonium thioglycolate ∼60% in water
Ammonium thioglycolate

CAS Number: 5421-46-5
MDL Number: MFCD00137451
Molecular Formula: C2H7NO2S
Molecular Weight: 109.14
Form: Clear Liquid
Color: Colorless


Other Product Names / Synonyms: Thioglycolic Acid Ammonium Salt, Ammonium Mercaptoacetate

Formula: C₂H₇NO₂S
MW: 109.14 g/mol
MDL Number: MFCD00137451
CAS Number: 5421-46-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=28fc629e927a71ad42f86d4fbda2408d7a79b73f&vid=85ec26d366eda1b2d5c8c03f04591c52fc9fb69e&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR