L-Thyroxine sodium salt pentahydrate ≥98.0%
Supplier: TCI America
Synonyms:
Sodium (S)-2-Amino-3-[4-(4-hydroxy-3,5-diiodophenoxy)-3,5-diiodophenyl]propionate pentahydrate, (S)-2-Amino-3-[4-(4-hydroxy-3,5-diiodophenoxy)-3,5-diiodophenyl]propionic acid sodium salt pentahydrate, Levothyroxine sodium pentahydrate
CAS Number: 55-03-8
MDL Number: MFCD00002596
Molecular Formula: C15H11I4NO4
Molecular Weight: 798.86
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: Very Pale Yellow
Specific rotation [a]20/D: 16 deg (C=1, 1mol/L HCL:EtOH=1:4)
Product Categories: Bioscience, Biologically-modified Amino Acids
Other Product Names / Synonyms: (S)-2-Amino-3-[4-(4-hydroxy-3,5-diiodophenoxy)-3,5-diiodophenyl]propionic Acid Sodium Salt, Sodium (S)-2-Amino-3-[4-(4-hydroxy-3,5-diiodophenoxy)-3,5-diiodophenyl]propionate
Formula:
C₁₅H₁₁I₄NO₄ MW: 798.86 g/mol |
MDL Number:
MFCD00002596 CAS Number: 55-03-8 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...