Monomethyl terephthalate ≥98.0%

Supplier: TCI America

Synonyms: Methyl hydrogen terephthalate, 4-(Methoxycarbonyl)benzoic acid

T0244-500G T0244-25G
TCT0244-500GEA 323 USD
TCT0244-500G TCT0244-025G
Monomethyl terephthalate ≥98.0%
Monomethyl terephthalate

CAS Number: 1679-64-7
MDL Number: MFCD00002557
Molecular Formula: C9H8O4
Molecular Weight: 180.16
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: White
Melting point (°C): 222


Other Product Names / Synonyms: Methyl Hydrogen Terephthalate, Terephthalic Acid Monomethyl Ester

Formula: C₉H₈O₄
MW: 180.16 g/mol
Melting Pt: 222 °C
MDL Number: MFCD00002557
CAS Number: 1679-64-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f40e93879d79be5b0c6ee5846e1cd64751b59e9e&vid=f40e93879d79be5b0c6ee5846e1cd64751b59e9e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR