Tetraiodo-2-sulfobenzoic anhydride ≥95.0% (by titrimetric analysis)

Supplier: TCI America

T0131-25G T0131-1G
TCT0131-025GEA 588 USD
TCT0131-025G TCT0131-001G
Tetraiodo-2-sulfobenzoic anhydride ≥95.0% (by titrimetric analysis)
Tetraiodo-2-sulfobenzoic anhydride

CAS Number: 1745-83-1
MDL Number: MFCD00059713
Molecular Formula: C7I4O4S
Molecular Weight: 687.75
Purity/Analysis Method: >95.0% (T)
Form: Crystal
Color: Yellow

Formula: C₇I₄O₄S
MW: 687.75 g/mol
MDL Number: MFCD00059713
CAS Number: 1745-83-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0f3e9a74381c6e2b4480b55336c5b276c639c0f0&vid=0f3e9a74381c6e2b4480b55336c5b276c639c0f0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR