Tetraethyl orthosilicate (TEOS) ≥97.0%
Supplier: TCI America
|
Danger
|
Synonyms:
Tetraethoxysilane
T0100-25ML
T0100-500ML
TCT0100-025MLEA
23
USD
TCT0100-025ML
TCT0100-500ML
Tetraethyl orthosilicate (TEOS) ≥97.0%
Tetraethyl orthosilicate (TEOS)
CAS Number: 78-10-4
MDL Number: MFCD00009062
Molecular Formula: C8H20O4Si
Molecular Weight: 208.33
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 165
Melting point (°C): -82
Flash Point (°C): 54
Specific Gravity (20/20): 0.93
Other Product Names / Synonyms: Tetraethoxysilane, TEOS
Formula:
Si(OC₂H₅)₄ MW: 208.33 g/mol Boiling Pt: 165 °C Melting Pt: -82 °C Density: 0.93 (20 °C) Flash Pt: 54 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009062 CAS Number: 78-10-4 EINECS: 201-083-8 UN: 1292 ADR: 3,III Merck Index: 13,03882 |
Specification Test Results
Purity/Analysis Method | >97.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...