α,α,α',α'-Tetrabromo-p-xylene ≥98.0%

Supplier: TCI America

T0053-25G T0053-5G
TCT0053-025GEA 482 USD
TCT0053-025G TCT0053-5G
α,α,α',α'-Tetrabromo-p-xylene ≥98.0%
α,α,α',α'-Tetrabromo-p-xylene

CAS Number: 1592-31-0
MDL Number: MFCD00017857
Molecular Formula: C8H6Br4
Molecular Weight: 421.75
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 170


Other Product Names / Synonyms: 1,4-Bis(dibromomethyl)benzene, p-Xylylene Tetrabromide

Formula: C₈H₆Br₄
Melting Pt: 170 °C
MDL Number: MFCD00017857
CAS Number: 1592-31-0
EINECS: 216-473-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=76c5ba705e1927c5e902c073c718c651766787f9&vid=92c81b2109fd6a6e12c80a40853fb711c8f31db2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR