α,α,α',α'-Tetrabromo-m-xylene ≥96.0%

Supplier: TCI America

T0051-25G T0051-10G
TCT0051-025GEA 159 USD
TCT0051-025G TCT0051-010G
α,α,α',α'-Tetrabromo-m-xylene ≥96.0%
α,α,α',α'-Tetrabromo-m-xylene

CAS Number: 36323-28-1
MDL Number: MFCD00000132
Molecular Formula: C8H6Br4
Molecular Weight: 421.75
Purity/Analysis Method: >96.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 106


Other Product Names / Synonyms: 1,3-Bis(dibromomethyl)benzene, m-Xylylene Tetrabromide

Formula: C₈H₆Br₄
Melting Pt: 106 °C
MDL Number: MFCD00000132
CAS Number: 36323-28-1
EINECS: 252-975-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d1eae3c6ac996b3cde1deefe2327a4df37789d6f&vid=d1eae3c6ac996b3cde1deefe2327a4df37789d6f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR