Tetrabromo-o-sulfobenzoic anhydride ≥98.0%

Supplier: TCI America

T0048-5G T0048-25G
TCT0048-005GEA 56 USD
TCT0048-005G TCT0048-025G
Tetrabromo-o-sulfobenzoic anhydride ≥98.0%
Tetrabromo-o-sulfobenzoic anhydride

CAS Number: 68460-01-5
MDL Number: MFCD00009745
Molecular Formula: C7Br4O4S
Molecular Weight: 499.75
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 220

Formula: C₇Br₄O₄S
MW: 499.75 g/mol
Melting Pt: 220 °C
MDL Number: MFCD00009745
CAS Number: 68460-01-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d459ae1ea3f5caf6632e9c01fd3d50fcea2e0bb8&vid=d459ae1ea3f5caf6632e9c01fd3d50fcea2e0bb8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR