1,13-Tridecanedioic acid ≥96.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: Tridecanedioic acid, Undecane-1,11-dicarboxylic acid, 1,11-Undecanedicarboxylic acid

T0021-25G T0021-5G
TCT0021-025GEA 144 USD
TCT0021-025G TCT0021-010G TCT0021-5G
1,13-Tridecanedioic acid ≥96.0% (by GC, titration analysis)
1,13-Tridecanedioic acid

CAS Number: 505-52-2
MDL Number: MFCD00002740
Molecular Formula: C13H24O4
Molecular Weight: 244.33
Purity/Analysis Method: >96.0% (GC,T)
Form: Crystal
Color: White
Melting point (°C): 113
Flash Point (°C): 236


Other Product Names / Synonyms: Brassylic Acid

Formula: C₁₃H₂₄O₄
MW: 244.33 g/mol
Boiling Pt: 230 °C (0.5 mmHg)
Melting Pt: 113 °C
Density: 1.15 g/cm³ (25 °C)
Flash Pt: 236 °C
Storage Temperature: Ambient
MDL Number: MFCD00002740
CAS Number: 505-52-2
EINECS: 208-011-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0b4473569e9304684ea4b6ce9b7eb8783fb1d8ce&vid=fbbfe8de67a95d3de5664193900aba9a69b278d2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >96.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR