1,1'-Sulfonyldiimidazole ≥98.0%

Supplier: TCI America

Synonyms: 1,1'-sulphonyldiimidazole

S0803-25G S0803-5G
TCS0803-25GEA 116 USD
TCS0803-25G TCS0803-5G
1,1'-Sulfonyldiimidazole ≥98.0%
1,1'-Sulfonyldiimidazole

CAS Number: 7189-69-7
MDL Number: MFCD00015893
Molecular Formula: C6H6N4O2S
Molecular Weight: 198.20
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 140

Formula: C₆H₆N₄O₂S
MW: 198.20 g/mol
Melting Pt: 140 °C
MDL Number: MFCD00015893
CAS Number: 7189-69-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9bdfd62f6976b6385088027c876ef3f051b1492c&vid=9bdfd62f6976b6385088027c876ef3f051b1492c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR