Silver sulfadiazine ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: Sulphadiazine Silver

S0595-25G S0595-5G
TCS0595-25GEA 188 USD
TCS0595-25G TCS0595-5G
Silver sulfadiazine ≥98.0% (by HPLC, titration analysis)
Silver sulphadiazine

CAS Number: 22199-08-2
MDL Number: MFCD00072101
Molecular Formula: C10H9AgN4O2S
Molecular Weight: 357.14
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Color: White
Melting point (°C): 285


Product Categories: Bioscience, Synthetic Antimicrobial Ingredients
Other Product Names / Synonyms: Sulfadiazine Silver

Formula: C₁₀H₉AgN₄O₂S
MW: 357.14 g/mol
Melting Pt: 285 °C
MDL Number: MFCD00072101
CAS Number: 22199-08-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=72f25f62ed247adf5132dc2bc272bb5438f79c12&vid=72f25f62ed247adf5132dc2bc272bb5438f79c12&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR