Stevioside ≥85.0% (by HPLC)
Supplier: TCI America
Synonyms:
(2S,3R,4S,5S,6R)-3,4,5-Trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl (4R,4aS,6aR,9S,11aR,11bS)-9-(((2S,3R,4S,5S,6R)-4,5-dihydroxy-6-(hydroxymethyl)-3-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)tetrahydro-2H-pyran-2-yl)oxy)-4,11b-dimethyl-8-methylenetetradecahydro-6a,9-methanocyclohepta[a]naphthalene-4-carboxylate
CAS Number: 57817-89-7
MDL Number: MFCD00079561
Molecular Formula: C38H60O18
Molecular Weight: 804.88
Purity/Analysis Method:
Form: Crystal
Color: White
Melting point (°C): 236
Specific rotation [a]20/D: -30 deg (C=6, H2O)
Formula:
C₃₈H₆₀O₁₈ MW: 804.88 g/mol Melting Pt: 236 °C |
MDL Number:
MFCD00079561 CAS Number: 57817-89-7 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...