Sulfamethazine sodium salt ≥97.0%

Supplier: TCI America

Synonyms: 4-Amino-N-(4,6-dimethyl-2-pyrimidyl)benzenesulphonamide sodium salt, Sodium ((4-aminophenyl)sulphonyl)(4,6-dimethylpyrimidin-2-yl)amide, Sulphadimidine sodium salt, Sodium sulphamethazine

S0583-25G S0583-500G
TCS0583-25GEA 37 USD
TCS0583-25G TCS0583-500G
Sulfamethazine sodium salt ≥97.0%
Sulfamethazine sodium salt

CAS Number: 1981-58-4
MDL Number: MFCD00068333
Molecular Formula: C12H14N4O2S
Molecular Weight: 300.31
Purity/Analysis Method: >97.0% (HPLC,N)
Form: Crystal
Color: White
Storage Temperature: 0-10°C


Product Categories: Bioscience, Synthetic Antimicrobial Ingredients
Other Product Names / Synonyms: Sodium Sulfamethazine, 4-Amino-N-(4,6-dimethyl-2-pyrimidyl)benzenesulfonamide Sodium Salt

Formula: C₁₂H₁₃N₄NaO₂S
MDL Number: MFCD00068333
CAS Number: 1981-58-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=90b1b88cb5de80a0cb06753e98d3d0406722f81e&vid=90b1b88cb5de80a0cb06753e98d3d0406722f81e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR