Sulfasalazine ≥95.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
5-[4-(2-Pyridylsulfamoyl)phenylazo]salicylic Acid
, 2-Hydroxy-5-((4-(N-(pyridin-2-yl)sulphamoyl)phenyl)diazenyl)benzoic acid
, 2-Hydroxy-5-((4-(N-(pyridin-2-yl)sulfamoyl)phenyl)diazenyl)benzoic acid
, 5-[4-(2-Pyridylsulphamoyl)phenylazo]salicylic Acid
CAS Number: 599-79-1
MDL Number: MFCD00057363
Molecular Formula: C18H14N4O5S
Molecular Weight: 398.39
Purity/Analysis Method:
Form: Crystal
Color: Yellow
Melting point (°C): 246
Product Categories: Bioscience, Immune System, Inflammation, Allergy Research Reagents, Immune System, Inflammation, Allergy Research Reagents, Inhibitors (for Pharmacological Research), Immunosuppressant Ingredients, Synthetic Antimicrobial Ingredients, Anti-rheumatoid Arth
Other Product Names / Synonyms: 5-[4-(2-Pyridylsulfamoyl)phenylazo]salicylic Acid
Formula:
C₁₈H₁₄N₄O₅S MW: 398.39 g/mol Melting Pt: 246 °C |
MDL Number:
MFCD00057363 CAS Number: 599-79-1 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...