Sulfacetamide sodium salt ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

S0578-250G S0578-25G
TCS0578-250GEA 163 USD
TCS0578-250G TCS0578-25G
Sulfacetamide sodium salt ≥98.0% (by HPLC, titration analysis)
Sulfacetamide sodium salt

CAS Number: 127-56-0
MDL Number: MFCD00007885
Molecular Formula: C8H10N2O3S
Molecular Weight: 236.22
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Color: White


Product Categories: Bioscience, Synthetic Antimicrobial Ingredients
Other Product Names / Synonyms: N-Acetyl-4-aminobenzenesulfonamide Sodium Salt, N-Acetylsulfanilamide Sodium Salt

Formula: C₈H₁₀N₂O₃S
MDL Number: MFCD00007885
CAS Number: 127-56-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=874b04fabd6d8f05aeeca92c286672bff7b93ab3&vid=874b04fabd6d8f05aeeca92c286672bff7b93ab3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR