Succinic acid dihydrazide ≥97.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
Succinic Dihydrazide
S0482-25G
S0482-500G
S0482-100G
TCS0482-25GEA
49
USD
TCS0482-25G
TCS0482-500G
TCS0482-100G
Succinic acid dihydrazide ≥97.0% (by HPLC, titration analysis)
Succinic acid dihydrazide
CAS Number: 4146-43-4
MDL Number: MFCD00007613
Molecular Formula: C4H10N4O2
Molecular Weight: 146.15
Purity/Analysis Method: >97.0% (HPLC,T)
Form: Crystal
Color: White
Melting point (°C): 168
Other Product Names / Synonyms: Butanediohydrazide
Formula:
C₄H₁₀N₄O₂ MW: 146.1478 g/mol Melting Pt: 168 °C |
MDL Number:
MFCD00007613 CAS Number: 4146-43-4 EINECS: 223-970-9 |
Specification Test Results
Purity/Analysis Method | >97.0% (HPLC,T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...