2,2'-sulfonylbis(4-tert-octylphenol) ≥96.0% (by GC, titration analysis)

Supplier: TCI America
Warning

S0478-5G S0478-1G
TCS0478-5GEA 381.8 USD
TCS0478-5G TCS0478-1G
2,2'-sulfonylbis(4-tert-octylphenol) ≥96.0% (by GC, titration analysis)
2,2'-sulfonylbis(4-tert-octylphenol)

CAS Number: 15452-89-8
MDL Number: MFCD00026328
Molecular Formula: C28H42O4S
Molecular Weight: 474.70
Purity/Analysis Method: >96.0% (GC,T)
Form: Crystal
Color: White
Melting point (°C): 148


Other Product Names / Synonyms: Bis(2-hydroxy-5-tert-octylphenyl) Sulfone, Bis[2-hydroxy-5-(1,1,3,3-tetramethylbutyl)phenyl] Sulfone

Formula: C₂₈H₄₂O₄S
Melting Pt: 148 °C
MDL Number: MFCD00026328
CAS Number: 15452-89-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2cfa481a1bc42cc1a2b9c7323200793ad3910367&vid=a24da83e23584f76876f2bb940778d9ab3136824&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 96.0 %
Purity (Neutralization titration) min. 96.0 %
Melting point 144.0 to 148.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR