L-Selenomethionine ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
L-SelenoMethionine, (S)-2-Amino-4-(methylseleno)butyric Acid, H-Se-Met-OH
S0442-1G
S0442-5G
TCS0442-001GEA
185.38
USD
TCS0442-001G
TCS0442-5G
L-Selenomethionine ≥98.0% (by HPLC, titration analysis)
L-Selenomethionine
CAS Number: 3211-76-5
MDL Number: MFCD00037210
Molecular Formula: C5H11NO2Se
Molecular Weight: 196.11
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Color: White
Melting point (°C): 267
Specific rotation [a]20/D: 18 deg (C=0.5, 2mol/L HCl)
Storage Temperature: <0°C
Product Categories: Bioscience, Antioxidants, Biologically-modified Amino Acids
Other Product Names / Synonyms: (S)-2-Amino-4-(methylseleno)butyric Acid
Formula:
C₅H₁₁NO₂Se MW: 196.11 g/mol Melting Pt: 267 °C |
MDL Number:
MFCD00037210 CAS Number: 3211-76-5 |
Specification Test Results
Purity/Analysis Method | >98.0% (HPLC,T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...