Stachyose hydrate ≥98.0% (by HPLC)
Supplier: TCI America
Synonyms:
Manneotetrose, beta-D-Fructofuranosyl O-alpha-D-Galactopyranosyl-(1-6)-O-alpha-D-galactopyranosyl-(1-6)-alpha-D-glucopyranoside, Stachyose hydrate(1:x), Lupeose
CAS Number: 54261-98-2
MDL Number: MFCD00071593
Molecular Formula: C24H42O21
Molecular Weight: 666.58
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Color: White
Melting point (°C): 110
Specific rotation [a]20/D: 133 deg (C=5, H2O)
Product Categories: Bioscience, Basic Sugars (Mono & Oligosaccharides), Tetrasaccharides
Other Product Names / Synonyms: beta-D-Fructofuranosyl O-alpha-D-Galactopyranosyl-(1-6)-O-alpha-D-galactopyranosyl-(1-6)-alpha-D-glucopyranoside, Lupeose, Manneotetrose
Formula:
C₂₄H₄₂O₂₁ MW: 666.58 g/mol (anhy) Melting Pt: 110 °C |
MDL Number:
MFCD00071593 CAS Number: 54261-98-2 |
Specification Test Results
Purity/Analysis Method | >98.0% (HPLC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...