trans-Stilbene oxide ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: trans-2,3-Diphenylethylene Oxide

S0262-25G S0262-5G
TCS0262-025GEA 326 USD
TCS0262-025G TCS0262-005G
trans-Stilbene oxide ≥98.0% (by HPLC)
trans-Stilbene oxide

CAS Number: 1439-07-2
MDL Number: MFCD00064311
Molecular Formula: C14H12O
Molecular Weight: 196.25
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Color: White
Melting point (°C): 68


Other Product Names / Synonyms: trans-2,3-Diphenylethylene Oxide

Formula: C₁₄H₁₂O
MW: 196.25 g/mol
Melting Pt: 68 °C
MDL Number: MFCD00064311
CAS Number: 1439-07-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4af1fcac24f5a5b461e112740ed97ea83cacfa83&vid=4af1fcac24f5a5b461e112740ed97ea83cacfa83&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR