Allyl phenoxyacetate ≥98.0%
Supplier: TCI America
Synonyms:
Phenoxyacetic Acid Allyl Ester
A2177-25G
A2177-250G
TCA2177-25GEA
49
USD
TCA2177-25G
TCA2177-250G
Allyl phenoxyacetate ≥98.0%
Allyl phenoxyacetate
CAS Number: 7493-74-5
MDL Number: MFCD00044632
Molecular Formula: C11H12O3
Molecular Weight: 192.21
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 137
Flash Point (°C): 113
Specific Gravity (20/20): 1.11
Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: Phenoxyacetic Acid Allyl Ester
Formula:
C₁₁H₁₂O₃ MW: 192.21 g/mol Boiling Pt: 137 °C Density: 1.11 (20 °C) Flash Pt: 113 °C |
MDL Number:
MFCD00044632 CAS Number: 7493-74-5 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...