Sebacic acid dihydrazide ≥96.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
Decanedihydrazide
S0224-25G
S0224-500G
TCS0224-025GEA
60
USD
TCS0224-025G
TCS0224-500G
Sebacic acid dihydrazide ≥96.0% (by titrimetric analysis)
Sebacic acid dihydrazide
CAS Number: 925-83-7
MDL Number: MFCD00039763
Molecular Formula: C10H22N4O2
Molecular Weight: 230.31
Purity/Analysis Method: >96.0% (T)
Form: Crystal
Color: White
Melting point (°C): 186
Other Product Names / Synonyms: Decanediohydrazide, Sebacohydrazide
Formula:
C₁₀H₂₂N₄O₂ MW: 230.3073 g/mol Melting Pt: 186 °C |
MDL Number:
MFCD00039763 CAS Number: 925-83-7 EINECS: 213-126-8 |
Specification Test Results
Purity/Analysis Method | >96.0% (T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...