Sulfonfluorescein ≥75.0% (by HPLC, titration analysis)

Supplier: TCI America

S0132-5G S0132-1G
TCS0132-5GEA 467.93 USD
TCS0132-5G TCS0132-001G
Sulfonfluorescein ≥75.0% (by HPLC, titration analysis)
Sulfonfluorescein

CAS Number: 4424-03-7 MDL Number: MFCD00070618 Molecular Formula: C19H12O6S Molecular Weight: 368.36 Purity/Analysis Method: 75.0% (HPLC,T) Form: Crystal Color: Deep Yellow Red

Formula: C₁₉H₁₂O₆S
MDL Number: MFCD00070618
CAS Number: 4424-03-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6aeacef84a311f4d54b1d95b9fe8df28067765a7&vid=6aeacef84a311f4d54b1d95b9fe8df28067765a7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 75.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR