Sodium sulfamate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

S0118-500G S0118-25G
TCS0118-500GEA 152 USD
TCS0118-500G TCS0118-025G
Sodium sulfamate ≥98.0% (by titrimetric analysis)
Sodium sulfamate

CAS Number: 13845-18-6
MDL Number: MFCD00059940
Molecular Formula: H3NO3S
Molecular Weight: 119.07
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White


Other Product Names / Synonyms: Aminosulfonic Acid Sodium Salt, Sulfamic Acid Sodium Salt

Formula: H₃NO₃S
MDL Number: MFCD00059940
CAS Number: 13845-18-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4c73aaa369e5b45e700f7bf76e743cf68d4425b8&vid=4c73aaa369e5b45e700f7bf76e743cf68d4425b8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR