Sucrose fatty acid ester
Supplier: TCI America
Synonyms:
Saccharose Fatty Acid Ester, Fatty Acid Sucrose Ester
S0112-500G
S0112-25G
TCS0112-500GEA
246.16
USD
TCS0112-500G
TCS0112-025G
Sucrose fatty acid ester
Sucrose fatty acid ester
MDL Number: MFCD00148531
Form: Crystal
Color: White
Other Product Names / Synonyms: Fatty Acid Sucrose Ester, Saccharose Fatty Acid Ester
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...