Spermine phosphate hexahydrate ≥98.0% (by total nitrogen basis)
Supplier: TCI America
Synonyms:
Neuridine Phosphate, Gerontine Phosphate, N,N'-Bis(3-aminopropyl)tetramethylenediamine Phosphate, Musculamine Phosphate, N,N'-Bis(3-aminopropyl)-1,4-butanediamine Phosphate
CAS Number: 58298-97-8
MDL Number: MFCD00050839
Molecular Formula: C10H26N4
Molecular Weight: 398.33
Purity/Analysis Method: >98.0% (N)
Form: Crystal
Color: White
Product Categories: Bioscience, Polyamines
Other Product Names / Synonyms: N,N'-Bis(3-aminopropyl)-1,4-butanediamine Phosphate, N,N'-Bis(3-aminopropyl)tetramethylenediamine Phosphate, Gerontine Phosphate, Musculamine Phosphate, Neuridine Phosphate
Formula:
C₁₀H₂₆N₄ MW: 398.33 g/mol |
MDL Number:
MFCD00050839 CAS Number: 58298-97-8 |
Specification Test Results
Purity/Analysis Method | >98.0% (N) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...