Span® 83

Supplier: TCI America

Synonyms: Arlacel C, Arlacel 83, Sorbitan Sesquioleate

S0063-25G S0063-500G
TCS0063-025GEA 20 USD
TCS0063-025G TCS0063-500G
Span® 83
Span® 83

CAS Number: 8007-43-0
MDL Number: MFCD00151568
Form: Clear Liquid
Color: Yellow
Flash Point (°C): 257
Specific Gravity (20/20): 0.99


Other Product Names / Synonyms: Arlacel 83, Arlacel C, Sorbitan Sesquioleate

Formula: C₄₂H₇₆O₇
MW: 693.06 g/mol
Density: 0.99 (20 °C)
Flash Pt: 257 °C
MDL Number: MFCD00151568
CAS Number: 8007-43-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR