Sodium sorbate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Sorbic Acid Sodium Salt

S0058-25G S0058-500G
TCS0058-025GEA 70 USD
TCS0058-025G TCS0058-500G
Sodium sorbate ≥98.0% (by titrimetric analysis)
Sodium sorbate

CAS Number: 7757-81-5
MDL Number: MFCD00058995
Molecular Formula: C6H8O2
Molecular Weight: 134.11
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White


Product Categories: Bioscience, Lipids
Other Product Names / Synonyms: Sorbic Acid Sodium Salt

Formula: C₆H₈O₂
MW: 134.11 g/mol
MDL Number: MFCD00058995
CAS Number: 7757-81-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dcbe0bf58b755d22a81e0204f6eb8ce0eb6d570a&vid=dcbe0bf58b755d22a81e0204f6eb8ce0eb6d570a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR