Scopolamine hydrobromide trihydrate ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

S0021-1G S0021-10G
TCS0021-001GEA 108.8 USD
TCS0021-001G TCS0021-010G
Scopolamine hydrobromide trihydrate ≥98.0% (by HPLC, titration analysis)
Scopolamine Hydrobromide Trihydrate

CAS Number: 6533-68-2 MDL Number: MFCD00012647 Molecular Formula: C17H21NO4 Molecular Weight: 384.27 Purity/Analysis Method: 98.0% (HPLC,T) Form: Crystal Color: White Melting point (°C): 195 Specific rotation [a]20/D: -25.5 deg (C=5, H2O)

Formula: C₁₇H₂₁NO₄
Melting Pt: 195 °C
MDL Number: MFCD00012647
CAS Number: 6533-68-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ab3ae3496a38251a72b56e0cb10b2643554340d4&vid=ab3ae3496a38251a72b56e0cb10b2643554340d4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR