Isobutyl salicylate ≥99.0%
Supplier: TCI America
Synonyms:
2-Hydroxybenzoic Acid Isobutyl Ester, Salicylic Acid Isobutyl Ester
S0013-25G
S0013-500G
TCS0013-025GEA
17
USD
TCS0013-025G
TCS0013-500G
Isobutyl salicylate ≥99.0%
Isobutyl salicylate
CAS Number: 87-19-4
MDL Number: MFCD00020035
Molecular Formula: C11H14O3
Molecular Weight: 194.23
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 259
Flash Point (°C): 121
Specific Gravity (20/20): 1.07
Other Product Names / Synonyms: 2-Hydroxybenzoic Acid Isobutyl Ester, Salicylic Acid Isobutyl Ester
Formula:
C₁₁H₁₄O₃ Boiling Pt: 259 °C Density: 1.07 (20 °C) Flash Pt: 121 °C |
MDL Number:
MFCD00020035 CAS Number: 87-19-4 |
|
Specification Test Results
Purity/Analysis Method | >99.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...