Ribavirin ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI America
Synonyms:
1-((2R,3R,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-1H-1,2,4-triazole-3-carboxamide
CAS Number: 36791-04-5
MDL Number: MFCD00058564
Molecular Formula: C8H12N4O5
Molecular Weight: 244.21
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Color: White
Specific rotation [a]20/D: -36 deg (C=1, H2O)
Product Categories: Bioscience, Antiviral Ingredients, Pharmacologically-Active Nucleosides and Nucleobases for Research and Experimental Use, Nucleosides and Their Analogs
Other Product Names / Synonyms: 1-(beta-D-Ribofuranosyl)-1,2,4-triazole-3-carboxamide
Formula:
C8H12N4O5 MW: 244.2047 g/mol |
MDL Number:
MFCD00058564 CAS Number: 36791-04-5 |
Specification Test Results
Purity/Analysis Method | >98.0% (HPLC,N) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...