Rhodium(II) acetate dimer

Supplier: TCI America

Synonyms: Dirhodium tetraacetate, Tetrakis(acetato)dirhodium(II)

R0069-100MG R0069-1G
TCR0069-100MGEA 164.2 USD
TCR0069-100MG TCR0069-1G
Rhodium(II) acetate dimer
Rhodium (II) acetate dimer

CAS Number: 15956-28-2
MDL Number: MFCD00003538
Molecular Formula: C8H12O8Rh2
Molecular Weight: 441.99
Form: Crystal
Color: Yellow Green
Melting point (°C): 240


Other Product Names / Synonyms: Dirhodium(II) Tetraacetate, Tetrakis(acetato)dirhodium(II)

Formula: C₈H₁₂O₈Rh₂
MW: 441.987 g/mol
Melting Pt: 240 °C
MDL Number: MFCD00003538
CAS Number: 15956-28-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e32d2314de76797726e632e31d96aecdf4a4ccae&vid=e32d2314de76797726e632e31d96aecdf4a4ccae&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR