D(+)-Ribonic acid γ-lactone ≥97.0%

Supplier: TCI America

Synonyms: (3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-one, Ribono-γ-lactone, D-Ribonic acid-1,4-lactone

R0063-1G R0063-5G
TCR0063-1GEA 64 USD
TCR0063-1G TCR0063-5G
D(+)-Ribonic acid γ-lactone ≥97.0%
D(+)-Ribonic acid γ-lactone

CAS Number: 5336-08-3
MDL Number: MFCD00063241
Molecular Formula: C5H8O5
Molecular Weight: 148.11
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 86
Specific rotation [a]20/D: 18 deg (C=1, H2O)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks, Bioscience, Sugar Acids, Ribose
Other Product Names / Synonyms: D-(+)-Ribonic Acid gamma-Lactone

Formula: C₅H₈O₅
MW: 148.11 g/mol
Melting Pt: 86 °C
MDL Number: MFCD00063241
CAS Number: 5336-08-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eea9889cd691bdb228da5527437be0f184801442&vid=eea9889cd691bdb228da5527437be0f184801442&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR