Amorolfine hydrochloride ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Amorolfin, Ro 14-4767/002, Amorolfine Hydrochloride, Curanail, Loceryl, (2R,6S)-rel-2,6-Dimethyl-4-(2-methyl-3-(4-(tert-pentyl)phenyl)propyl)morpholine hydrochloride, (2R,6S)-rel-4-[3-[4-(1,1-Dimethylpropyl)phenyl]-2-methylpropyl]-2,6-dimethyl-morpholine hydrochloride

A2161-1G A2161-200MG
TCA2161-1GEA 223.9 USD
TCA2161-1G TCA2161-200MG
Amorolfine hydrochloride ≥98.0% (by titrimetric analysis)
Amorolfine hydrochloride

CAS Number: 78613-38-4
MDL Number: MFCD00903738
Molecular Formula: C21H35NO
Molecular Weight: 353.98
Purity/Analysis Method: >98.0% (T)
Form: Crystal

Formula: C₂₁H₃₅NO
MW: 353.98 g/mol
MDL Number: MFCD00903738
CAS Number: 78613-38-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dda98b77dd3a9e93ced80012ae13f1b0e07154e0&vid=dda98b77dd3a9e93ced80012ae13f1b0e07154e0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR