L(+)-Rhamnose monohydrate ≥98.0% (by HPLC)

Supplier: TCI America

R0013-5G R0013-25G
TCR0013-005GEA 32.98 USD
TCR0013-005G TCR0013-025G
L(+)-Rhamnose monohydrate ≥98.0% (by HPLC)
L(+)-Rhamnose monohydrate

CAS Number: 10030-85-0
MDL Number: MFCD00149363
Molecular Formula: C6H12O5
Molecular Weight: 164.16
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Color: White
Melting point (°C): 93
Specific rotation [a]20/D: 8 deg (C=10, H2O)


Product Categories: Bioscience, Rhamnose, Deoxysugars
Other Product Names / Synonyms: 6-Deoxy-L-mannose

Formula: C₆H₁₂O₅·1H₂O
Melting Pt: 93 °C
Density: 1.47 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149363
CAS Number: 10030-85-0
EINECS: 222-793-4
Merck Index: 12,08338
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=98ba69487977aa7c90c16eab38619755bae883c1&vid=98ba69487977aa7c90c16eab38619755bae883c1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR