Quinoline-N-oxide hydrate ≥97.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Quinoline 1-oxide hydrate

Q0072-5G Q0072-25G
TCQ0072-5GEA 144 USD
TCQ0072-5G TCQ0072-25G
Quinoline-N-oxide hydrate ≥97.0% (by titrimetric analysis)
Quinoline-N-oxide hydrate

CAS Number: 198878-42-1
Molecular Formula: C9H7NO
Molecular Weight: 145.16
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Color: White
Boiling point (°C): 173
Melting point (°C): 59

Formula: C₉H₇NO·xH₂O
MW: 145.16 g/mol (anhy)
Boiling Pt: 173 °C
Melting Pt: 59 °C
MDL Number: MFCD00149472
CAS Number: 64201-64-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fd8092611f72311b286b62e041f5ae6c339237d4&vid=fd8092611f72311b286b62e041f5ae6c339237d4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR