Quinolinium dichromate ≥97.0%
Supplier: TCI America
Q0058-10G
Q0058-25G
TCQ0058-010GEA
76
USD
TCQ0058-010G
TCQ0058-25G
Quinolinium dichromate ≥97.0%
Quinolinium dichromate
CAS Number: 56549-24-7
MDL Number: MFCD00059108
Molecular Formula: C18H16Cr2N2O7
Molecular Weight: 476.33
Purity/Analysis Method:
Formula:
C₁₈H₁₆Cr₂N₂O₇ MW: 476.33 g/mol Melting Pt: 166 °C |
MDL Number:
MFCD00059108 CAS Number: 56549-24-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...