6-Amyl-2-pyrone ≥96.0%
Supplier: TCI America
CAS Number: 27593-23-3
MDL Number: MFCD00047551
Molecular Formula: C10H14O2
Molecular Weight: 166.22
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 93
Flash Point (°C): 146
Specific Gravity (20/20): 1.01
Other Product Names / Synonyms: 6-Pentyl-2-pyrone, 6-Pentyl-2H-pyran-2-one
Formula:
C₁₀H₁₄O₂ Boiling Pt: 93 °C Density: 1.01 (20 °C) Flash Pt: 146 °C |
MDL Number:
MFCD00047551 CAS Number: 27593-23-3 |
|
Specification Test Results
Purity/Analysis Method | >96.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...