p-Quinone-4-chloroimide ≥95.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: N-Chloro-p-benzoquinone Monoimine, N-Chloro-p-quinone Monoimine

Q0019-5G Q0019-1G
TCQ0019-005GEA 328 USD
TCQ0019-005G TCQ0019-001G
p-Quinone-4-chloroimide ≥95.0% (by titrimetric analysis)
p-Quinone-4-chloroimide

CAS Number: 637-61-6
MDL Number: MFCD00058961
Molecular Formula: C6H4ClNO
Molecular Weight: 141.55
Purity/Analysis Method: >95.0% (T)
Form: Crystal
Color: Pale Yellow
Melting point (°C): 85


Other Product Names / Synonyms: N-Chloro-p-benzoquinone Monoimine, N-Chloro-p-quinone Monoimine

Formula: C₆H₄ClNO
MW: 141.55 g/mol
Melting Pt: 85 °C
MDL Number: MFCD00058961
CAS Number: 637-61-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5392153ac643e5216fdf92079595f2410e2bb058&vid=5392153ac643e5216fdf92079595f2410e2bb058&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR