Quinidine sulfate Dihydrate ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
(S)-(6-methoxyquinolin-4-yl)((1S,2R,4S,5R)-5-vinylquinuclidin-2-yl)methanol sulphate hydrate(2:1:2)
Q0010-5G
Q0010-25G
TCQ0010-005GEA
59
USD
TCQ0010-005G
TCQ0010-025G
Quinidine sulfate Dihydrate ≥98.0% (by titrimetric analysis)
Quinidine sulfate Dihydrate
CAS Number: 6591-63-5
MDL Number: MFCD00135926
Molecular Formula: C20H24N2O2
Molecular Weight: 746.92
Purity/Analysis Method:
Form: Crystal
Color: White
Melting point (°C): 212
Specific rotation [a]20/D: 275 deg (C=2, 0.1mol/L HCl)
Formula:
C₄₀H₄₈N₄O₄.H₂O₄S·2H₂O MW: 782.94 g/mol Melting Pt: 212 °C |
MDL Number:
MFCD00149346 CAS Number: 6591-63-5 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...