2-Phosphonobutane-1,2,4-tricarboxylic acid ∼50% in water

Supplier: TCI America

P1857-25G P1857-500G
TCP1857-25GEA 34 USD
TCP1857-25G TCP1857-500G
2-Phosphonobutane-1,2,4-tricarboxylic acid ∼50% in water
2-Phosphonobutane-1,2,4-tricarboxylic acid

CAS Number: 37971-36-1
MDL Number: MFCD01940753
Molecular Formula: C7H11O9P
Molecular Weight: 270.13
Form: Clear Liquid
Color: Colorless


Other Product Names / Synonyms: 3-Carboxy-3-phosphonohexanedioic Acid, PBTC

Formula: C₇H₁₁O₉P
MDL Number: MFCD01940753
CAS Number: 37971-36-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f7822c405b0f92e490b3541c2619d07d178fd6c1&vid=a15f50e07f87fed2eaf3ea4b5ab977aba199af90&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR