Pravastatin sodium ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
(3R,5R)-3,5-Dihydroxy-7-((1S,2S,6S,8S,8aR)-6-hydroxy-2-methyl-8-(((S)-2-methylbutanoyl)oxy)-1,2,6,7,8,8a-hexahydronaphthalen-1-yl)heptanoic acid, sodium salt
P1796-500MG
P1796-100MG
TCP1796-500MGEA
997
USD
TCP1796-500MG
TCP1796-100MG
Pravastatin sodium ≥98.0% (by HPLC, titration analysis)
Pravastatin sodium
CAS Number: 81131-70-6
MDL Number: MFCD00887601
Molecular Formula: C23H36O7
Molecular Weight: 446.52
Purity/Analysis Method:
Formula:
C₂₃H₃₅NaO₇ MW: 446.52 g/mol Melting Pt: 173 °C |
MDL Number:
MFCD00887601 CAS Number: 81131-70-6 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...