N-Phthaloyl-DL-glutamic anhydride ≥98.0%

Supplier: TCI America

Synonyms: 2-Phthalimidoglutaric Anhydride

P1789-25G P1789-5G
TCP1789-25GEA 444 USD
TCP1789-25G TCP1789-5G
N-Phthaloyl-DL-glutamic anhydride ≥98.0%
N-Phthaloyl-DL-glutamic anhydride

CAS Number: 3343-28-0
MDL Number: MFCD00005886
Molecular Formula: C13H9NO5
Molecular Weight: 259.22
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Melting point (°C): 205


Other Product Names / Synonyms: 2-Phthalimidoglutaric Anhydride

Formula: C₁₃H₉NO₅
Melting Pt: 205 °C
MDL Number: MFCD00005886
CAS Number: 3343-28-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=591a166b4d4aa85e86381f9626804453018ea5af&vid=591a166b4d4aa85e86381f9626804453018ea5af&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR