(S)-3-(4-Aminophenyl)-2-((tert-butoxycarbonyl)amino)propanoic acid ≥96.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 4-Amino-N-Boc-L-phenylalanine, Boc-p-amino-Phe-OH, Boc-Phe(4-NH2)-OH

A2131-1G A2131-5G
TCA2131-1GEA 62.8 USD
TCA2131-1G TCA2131-5G
(S)-3-(4-Aminophenyl)-2-((tert-butoxycarbonyl)amino)propanoic acid ≥96.0% (by HPLC, titration analysis)
(S)-3-(4-Aminophenyl)-2-((tert-butoxycarbonyl)amino)propanoic acid

CAS Number: 55533-24-9
MDL Number: MFCD00038139
Molecular Formula: C14H20N2O4
Molecular Weight: 280.32
Purity/Analysis Method: >96.0% (HPLC,T)
Form: Crystal
Melting point (°C): 126
Specific rotation [a]20/D: 26 deg (C=1, MeOH)


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: 4-Amino-N-Boc-L-phenylalanine, Boc-Phe(4-NH2)-OH

Formula: C₁₄H₂₀N₂O₄
MW: 280.3 g/mol
Melting Pt: 126 °C
MDL Number: MFCD00038139
CAS Number: 55533-24-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8e0f60768f7c00e3e9c6325997462bd0bf97013e&vid=8e0f60768f7c00e3e9c6325997462bd0bf97013e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >96.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR