Potassium tris(3,5-dimethylpyrazol-1-yl)borohydride ≥98.0%

Supplier: TCI America

Synonyms: Potassium Hydrotris(3,5-dimethylpyrazol-1-yl)borate, Potassium hydrotris(3,5-dimethylpyrazol-1-yl)borate

P1441-25G P1441-5G
TCP1441-25GEA 422.8 USD
TCP1441-25G TCP1441-5G
Potassium tris(3,5-dimethylpyrazol-1-yl)borohydride ≥98.0%
Potassium tris(3,5-dimethylpyrazol-1-yl)borohydride

CAS Number: 17567-17-8
MDL Number: MFCD00040342
Molecular Formula: C15H22BKN6
Molecular Weight: 336.29
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White


Other Product Names / Synonyms: Potassium Hydrotris(3,5-dimethylpyrazol-1-yl)borate

Formula: C₁₅H₂₂BKN₆
MW: 336.29 g/mol
MDL Number: MFCD00040342
CAS Number: 17567-17-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=92332a21415a92ae383ec4816383347b4454878a&vid=92332a21415a92ae383ec4816383347b4454878a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR