3A-Amino-3A-deoxy-(2AS,3AS)-ɑ-cyclodextrin hydrate ≥90.0% (by HPLC)
Supplier: TCI America
Synonyms:
3A-Amino-3A-deoxy-(2AS,3AS)-α-cyclodextrin Hydrate
A2122-1G
A2122-200MG
TCA2122-1GEA
317.79
USD
TCA2122-1G
TCA2122-200MG
3A-Amino-3A-deoxy-(2AS,3AS)-ɑ-cyclodextrin hydrate ≥90.0% (by HPLC)
3A-Amino-3A-deoxy-(2AS,3AS)-alpha-cyclodextrin hydrate
CAS Number: 121916-94-7
Molecular Formula: C36H61NO29
Molecular Weight: 971.86
Purity/Analysis Method: >90.0% (HPLC)
Form: Crystal
Specific rotation [a]20/D: 132.5 deg (C=1, H2O)
Formula:
C₃₆H₆₁NO₂₉ MW: 971.86 g/mol |
CAS Number:
121916-94-7 |
Specification Test Results
Purity/Analysis Method | >90.0% (HPLC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...