Potassium bis(1-pyrazolyl)borohydride ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Potassium Dihydrobis(1-pyrazolyl)borate

P1439-1G P1439-5G
TCP1439-1GEA 101 USD
TCP1439-1G TCP1439-5G
Potassium bis(1-pyrazolyl)borohydride ≥98.0% (by titrimetric analysis)
Potassium bis(1-pyrazolyl)borohydride

CAS Number: 18583-59-0
MDL Number: MFCD00058739
Molecular Formula: C6H8BKN4
Molecular Weight: 186.07
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Melting point (°C): 168


Other Product Names / Synonyms: Potassium Dihydrobis(1-pyrazolyl)borate

Formula: C₆H₈BKN₄
MW: 186.07 g/mol
Melting Pt: 168 °C
MDL Number: MFCD00058739
CAS Number: 18583-59-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4fd0da63e3bc2831cba6b573e120e83752b32812&vid=4fd0da63e3bc2831cba6b573e120e83752b32812&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR