Pentaethylene glycol monobenzyl ether ≥95.0%
Supplier: TCI America
Synonyms:
BnO-PEG5-OH, 2-[2-[2-[2-[2-(Benzyloxy)ethoxy]ethoxy]ethoxy]ethoxy]ethanol, 1-Phenyl-2,5,8,11,14-pentaoxahexadecan-16-ol
P1435-5G
P1435-1G
TCP1435-5GEA
682.1
USD
TCP1435-5G
TCP1435-1G
Pentaethylene glycol monobenzyl ether ≥95.0%
Pentaethylene glycol monobenzyl ether
CAS Number: 57671-28-0
MDL Number: MFCD06797154
Molecular Formula: C17H28O6
Molecular Weight: 328.41
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Color: Very Pale Yellow
Specific Gravity (20/20): 1.10
Other Product Names / Synonyms: 2-[2-[2-[2-[2-(Benzyloxy)ethoxy]ethoxy]ethoxy]ethoxy]ethanol
Formula:
C₁₇H₂₈O₆ Density: 1.10 (20 °C) |
MDL Number:
MFCD06797154 CAS Number: 57671-28-0 |
|
Specification Test Results
Purity/Analysis Method | >95.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...