(±)-2-Piperazinecarboxylic acid dihydrochloride
Supplier: TCI America
Synonyms:
Piperazine-2-carboxylic acid dihydrochloride
P1391-25G
P1391-5G
TCP1391-25GEA
151
USD
TCP1391-25G
TCP1391-5G
(±)-2-Piperazinecarboxylic acid dihydrochloride
(±)-2-Piperazinecarboxylic acid dihydrochloride
CAS Number: 3022-15-9
MDL Number: MFCD00044803
Molecular Formula: C5H10N2O2
Molecular Weight: 203.06
Form: Crystal
Color: White
Formula:
C₅H₁₂Cl₂N₂O₂ MW: 203.07 g/mol |
MDL Number:
MFCD00044803 CAS Number: 3022-15-9 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...